Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.85%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, natural, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, tablet
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing