Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride, AnhydroBeads™, −10 mesh, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride, AnhydroBeads™, −10 mesh, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%
Sign Into View Organizational & Contract Pricing