MilliporeSigma
Acrylamide levels in selected Colombian foods.
Food additives & contaminants. Part B, Surveillance (2014-12-17)
Deborah Pacetti, Elizabeth Gil, Natale G Frega, Lina Álvarez, Pilar Dueñas, Angélica Garzón, Paolo Lucci
ABSTRACT
Acrylamide (AA) levels in conventional (n = 112) and traditional (n = 43) Colombian foods were analysed by gas chromatography with mass spectrometry (GC/MS) detection. Samples included: infant powdered formula, coffee and chocolate powders, corn snacks, bakery products and tuber-, meat- and vegetable-based foods. There was a wide variability in AA levels among different foods and within different brands of the same food, especially for coffee powder, breakfast cereals biscuits and French fries samples. Among the conventional foods tested, the highest mean AA value was found in bakery products, such as biscuit (1104 µg kg(-1)) and wafer (1449 µg kg(-1)), followed by potato chips (916 µg kg(-1)). On the other hand, among the traditional foods, higher AA amounts were detected in fried platano (2813 µg kg(-1)) and yuca (3755 µg kg(-1)) compared to other products. Interestingly, the arepa, a traditional Colombian bakery product made with corn flour, showed a lower AA content (< 75 µg kg(-1)) when compared with similar bakery products tested, such as soft bread (102-594 µg kg(-1)), which is a made with wheat flour.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Heptane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Heptane, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Heptane, HPLC grade, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC