MilliporeSigma
  • Endocannabinoid and nitric oxide interaction mediates food intake in neonatal chicken.
Endocannabinoid and nitric oxide interaction mediates food intake in neonatal chicken.
British poultry science (2015-06-09)
S Hassanpour, M Zendehdel, V Babapour, S Charkhkar
ABSTRACT
The aim of the current study was to investigate the interaction of the nitric oxide and cannabinoidergic systems on feeding behaviour in neonatal chicken. A total of 6 experiments were designed to evaluate the interaction between cannabinoidergic and nitrergic systems on food intake in 3-h food-deprived (FD3) neonatal chickens. In Experiment 1, chickens received intracerebroventricular (ICV) injections of saline, 2-arachidonoylglycerol (2-AG) (a CB1 receptor agonist, 2 µg), l-arginine (nitric oxide precursor, 200 nmol) and co-administration of 2-AG + l-arginine. In Experiment 2, ICV injection of saline, 2-AG (2 µg), l-NAME (a nitric oxide synthesis inhibitor, 100 nmol) and their combination (2-AG + l-NAME) were applied to the birds. In Experiment 3, injections were saline, CB65 (a CB2 receptor agonist, 1.25 µg), l-arginine (200 nmol) and CB65 + l-arginine. In Experiment 4, birds received ICV injection of saline, CB65 (1.25 µg), l-NAME (100 nmol) and CB65 + l-NAME. In Experiment 5, chickens were ICV injected with saline, l-arginine (800 nmol), SR141716A (a selective CB1 receptor antagonist, 6.25 µg) and l-arginine + SR141716A. In Experiment 6, birds were injected with saline, l-arginine (800 nmol), AM630 (a selective CB2 receptor antagonist, 5 µg) and l-arginine + AM630. Cumulative food intake was recorded until 2-h post injection. ICV injection of CB1 and CB2 receptor agonists increased food intake. Co-injection of 2-AG + l-NAME increased the hyperphagic effects of CB1 receptors. CB2 receptor-induced food intake was not affected by co-administration of CB65 + l-NAME. l-Arginine decreased food intake and this effect was amplified by co-injection of l-arginine + SR141716A. However; CB2 receptor antagonists had no effect on l-arginine-induced hypophagia. The results suggest that there is an interaction between endogenous nitric oxide and the cannabinoidergic system on feeding behaviour which is mediated via CB1 receptors in the neonatal chicken.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C