MilliporeSigma
  • Dimethyl Fumarate Protects Neural Stem/Progenitor Cells and Neurons from Oxidative Damage through Nrf2-ERK1/2 MAPK Pathway.
Dimethyl Fumarate Protects Neural Stem/Progenitor Cells and Neurons from Oxidative Damage through Nrf2-ERK1/2 MAPK Pathway.
International journal of molecular sciences (2015-06-20)
Qin Wang, Sergei Chuikov, Sophina Taitano, Qi Wu, Arjun Rastogi, Samuel J Tuck, Joseph M Corey, Steven K Lundy, Yang Mao-Draayer
ABSTRACT
Multiple sclerosis (MS) is the most common multifocal inflammatory demyelinating disease of the central nervous system (CNS). Due to the progressive neurodegenerative nature of MS, developing treatments that exhibit direct neuroprotective effects are needed. Tecfidera™ (BG-12) is an oral formulation of the fumaric acid esters (FAE), containing the active metabolite dimethyl fumarate (DMF). Although BG-12 showed remarkable efficacy in lowering relapse rates in clinical trials, its mechanism of action in MS is not yet well understood. In this study, we reported the potential neuroprotective effects of dimethyl fumarate (DMF) on mouse and rat neural stem/progenitor cells (NPCs) and neurons. We found that DMF increased the frequency of the multipotent neurospheres and the survival of NPCs following oxidative stress with hydrogen peroxide (H2O2) treatment. In addition, utilizing the reactive oxygen species (ROS) assay, we showed that DMF reduced ROS production induced by H2O2. DMF also decreased oxidative stress-induced apoptosis. Using motor neuron survival assay, DMF significantly promoted survival of motor neurons under oxidative stress. We further analyzed the expression of oxidative stress-induced genes in the NPC cultures and showed that DMF increased the expression of transcription factor nuclear factor-erythroid 2-related factor 2 (Nrf2) at both levels of RNA and protein. Furthermore, we demonstrated the involvement of Nrf2-ERK1/2 MAPK pathway in DMF-mediated neuroprotection. Finally, we utilized SuperArray gene screen technology to identify additional anti-oxidative stress genes (Gstp1, Sod2, Nqo1, Srxn1, Fth1). Our data suggests that analysis of anti-oxidative stress mechanisms may yield further insights into new targets for treatment of multiple sclerosis (MS).
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
Dimethyl fumarate, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00
Sigma-Aldrich
DAPI, for nucleic acid staining
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
Only 2 left in stock
Details...
$336.00
1 g
In Stock
Details...
$596.00