MilliporeSigma
  • Functional characterisation and expression analysis of recombinant serum amyloid P isoform 1 (RbSAP1) from rock bream (Oplegnathus fasciatus).
Functional characterisation and expression analysis of recombinant serum amyloid P isoform 1 (RbSAP1) from rock bream (Oplegnathus fasciatus).
Fish & shellfish immunology (2015-04-29)
Kwang-Min Choi, Sang Hee Shim, Cheul Min An, Bo-Hye Nam, Ji-Min Jeong, Ju-Won Kim, Chan-Il Park
ABSTRACT
Lectins are carbohydrate-binding proteins that play important roles in the recognition and elimination of pathogens via the innate immune system. Pentraxins (PTX) are humoral lectins, which are multifunctional proteins in vertebrates. Pentraxins can be divided into two groups based on their primary structure: short (C-reactive protein and serum amyloid P [SAP]) and long pentraxins (PTX3 and neuronal pentraxins). Previously, SAP was shown to have Ca(2+)-dependent binding specificity for various ligands and to be a major acute phase protein. In this study, we identified and characterised the gene encoding SAP isoform 1 in rock bream (Oplegnathus fasciatus) (RbSAP1) and analysed its expression in various tissues after a pathogen challenge. An alignment analysis conducted based on the deduced amino acid sequence of RbSAP1 (1918 bp full-length cDNA with a 699 bp open reading frame encoding 232 amino acids) and SAPs and PTXs isolated from other organisms, revealed that the pentraxin domain and cysteine residues of the deduced protein are conserved. RbSAP1, which was ubiquitously expressed in all tissues examined, was predominantly detected in head kidney, trunk kidney, peripheral blood leukocytes, and gills. RbSAP1 expression was dramatically up-regulated in the kidney and liver after infection with Edwardsiella tarda, Streptococcus iniae, or red seabream iridovirus. Purified rRbSAP1 was able to bind Gram-negative bacteria, Gram-positive bacteria, and pathogen-associated molecular patterns. Interestingly, rRbSAP1 aggregated Gram-negative bacteria in the presence of Ca(2+). The anti-pathogen activity of rRbSAP1 suggests that SAP functions in innate immunity in the rock bream.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%