Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ACS reagent
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Calcium chloride
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sign Into View Organizational & Contract Pricing