MilliporeSigma
  • Quantitation of Synthetic Cannabinoids in Plant Materials Using High Performance Liquid Chromatography with UV Detection (Validated Method).
Quantitation of Synthetic Cannabinoids in Plant Materials Using High Performance Liquid Chromatography with UV Detection (Validated Method).
Journal of forensic sciences (2015-07-16)
Laura A Ciolino
ABSTRACT
Plant based products laced with synthetic cannabinoids have become popular substances of abuse over the last decade. Quantitative analysis for synthetic cannabinoid content in the laced materials is necessary for health hazard assessments addressing overall exposure and toxicity when the products are smoked. A validated, broadly applicable HPLC-UV method for the determination of synthetic cannabinoids in plant materials is presented, using acetonitrile extraction and separation on a commercial phenylhexyl stationary phase. UV detection provides excellent sensitivity with limits of quantitation (LOQs) less than 10 μg/g for many cannabinoids. The method was validated for several structural classes (dibenzopyrans, cyclohexylphenols, naphthoylindoles, benzoylindoles, phenylacetylindoles, tetramethylcyclopropylindoles) based on spike recovery experiments in multiple plant materials over a wide cannabinoid contents range (0.1-81 mg/g). Average recovery across 32 cannabinoids was 94% for marshmallow leaf, 95% for damiana leaf, and 92% for mullein leaf. The method was applied to a series of case-related products with determined amounts ranging from 0.2 to >100 mg/g.
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Triethylamine, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Triethylamine, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, ≥99.7%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, ≥99.7%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00