MilliporeSigma
  • Effects of Stirring and Fluid Perfusion on the In Vitro Degradation of Calcium Phosphate Cement/PLGA Composites.
Effects of Stirring and Fluid Perfusion on the In Vitro Degradation of Calcium Phosphate Cement/PLGA Composites.
Tissue engineering. Part C, Methods (2015-06-23)
Jie An, Sander C G Leeuwenburgh, Joop G C Wolke, John A Jansen
ABSTRACT
In vitro degradation rates of calcium phosphate bioceramics are investigated using a large variation of soaking protocols that do not all match the dynamic conditions of the perfused physiological environment. Therefore, we studied the effect of stirring and fluid perfusion on the in vitro degradation rate of apatitic calcium phosphate cements (CPC) containing poly(lactic-co-glycolic acid) (PLGA) microspheres. The composites were soaked in phosphate-buffered saline up to 6 weeks under unstirred, stirred, or perfused conditions followed by analysis of mass loss, compression strength, porosity, crystal phase composition, and morphology of the cement composites. The results showed that fluid perfusion reduced the decrease in pH and corresponding degradation rates, while nonperfused soaking conditions (i.e., stirred and unstirred conditions) resulted into more extensive acidification, the rate of which increased with stirring. After 2 weeks, the formation of a secondary brushite phase was observed for cement composites soaked under nonperfused (i.e., stirred and unstirred) conditions, whereas this phase was not detected in cements soaked under perfused conditions. The degradation rate of cement composites decreased in the order unstirred>stirred>perfused, as evidenced by quantification of mass loss, compression strength, and pore morphology. To summarize, we have demonstrated that soaking conditions strongly affected the in vitro degradation process of CPCs. As a consequence, it can be concluded that the experimental design of current in vitro degradation studies does not allow for correlation to (pre-)clinical studies.
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
2-Propanol, suitable for HPLC
Sigma-Aldrich
DL-Lactic acid, JIS special grade, 85.0-92.0%
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
2-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)