MilliporeSigma
  • Oncostatin M protects against demyelination by inducing a protective microglial phenotype.
Oncostatin M protects against demyelination by inducing a protective microglial phenotype.
Glia (2015-04-30)
Kris Janssens, Anurag Maheshwari, Chris Van den Haute, Veerle Baekelandt, Piet Stinissen, Jerome J A Hendriks, Helena Slaets, Niels Hellings
ABSTRACT
Multiple sclerosis (MS) is a chronic disabling disease of the central nervous system (CNS), in which destruction of myelin sheaths leads to disturbed axonal conduction. Available MS therapies modulate the immune response, but are unable to prevent neurological decline. Therefore, great efforts are made to develop therapies that limit demyelination and axonal degeneration. Oncostatin M (OSM), a member of the interleukin (IL)-6 cytokine family, is produced in demyelinating lesions of MS patients and stimulates neuronal survival. In this study, we reveal that the OSM receptor (OSMR) was robustly upregulated on microglia/macrophages and astrocytes in the cuprizone-induced demyelination model. While OSMR deficiency led to aggravated demyelination, CNS-targeted OSM treatment largely prevented demyelination. OSM treatment increased IL-4 expression and induced polarization of myeloid cells towards an anti-inflammatory M2 phenotype in vivo. This study reveals a previously uncharacterized and protective role for OSM during demyelination, and indicates that OSM is a promising therapeutic candidate to limit CNS damage in demyelinating diseases including MS.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, natural, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%