GC Analysis of Blood Alcohols on a Packed Column (5% Carbowax® 20M on 60/80 Carbopack™ B)
Materials
related product
Product No.
Description
Pricing
CONDITIONS
column
6 ft.x 2 mm I.D. glass
packing
5% Carbowax 20M on 60/80 Carbopack B
oven
85 °C
detector
FID
carrier gas
helium, 20 mL/min
injection
1 μL
sample
0.05-0.10% each analyte in water
Description
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Carbowax is a registered trademark of Union Carbide