Milli-Q® Ultrapure Water Solutions
Discover our expanded ultrapure system offer, designed for modern-day scientists
FIND THE PERFECT Type 1 WATER SYSTEM FOR YOUR LAB
You now have more choice of Milli-Q® ultrapure (Type 1) systems to perfectly satisfy your laboratory needs — and constraints.
- Choose from our state-of-the-art Milli-Q® IQ systems that offer a full Qualification program, advanced data traceability, and accurate TOC monitoring. Identified as Greener Alternative Products, these systems are certified to use less water, electricity, materials and packaging, and are completely mercury free (review the Design for Sustainability scorecards).
- Or discover our Milli-Q® EQ systems with their intuitive touchscreens, agile Q-POD® dispensing, and space-saving installation options. These systems offer dedicated innovations for flexibility to adapt to your lab’s constraints.
CONFIDENCE IN HIGH-QUALITY MILLI-Q® Analytical Grade lab WATER
With any Milli-Q® ultrapure water system, you can be sure that every dispense is high quality. Both Milli-Q® IQ and EQ 7 series systems contain the same powerful combination of purification media to provide ultrapure water quality consistently to the trace level. You can fully rely on these systems to deliver consistent water quality for your sensitive experiments, no matter your feed water quality. Plus, both types of systems can be connected to MyMilli-Q™ Remote Care monitoring and service capability, to assure productivity and quality.
Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Water System
The Ultimate Lab Water Solution
Exceed your most demanding expectations – from improved productivity to reduced environmental impact.
Milli-Q® IQ 7000 Ultrapure Water System
Invest in Lab Efficiency
See how the Milli-Q® IQ 7000 system can change the way you work in the lab.
Milli-Q® EQ 7008/16 Ultrapure & Pure Water System
Flexibility that Adapts to Your Lab
The authentic Milli-Q® ultrapure experience, designed for modern-day scientists.
Four Lab Water SYSTEMS. ONE AUTHENTIC MILLI-Q® EXPERIENCE.
- Easy and agile Q-POD® dispensing. Dispensing ultrapure water is easy, precise, and effortless with the “thumb wheel”. Access a range of dispensing functions that are useful for day-to-day work in the laboratory.
- Intuitive touchscreen control. Go anywhere in just a few clicks. The interactive screen simplifies system use and data access.
- Space-saving design. Only interact with the POD dispenser and touchscreen -- these compact systems can easily be integrated into any lab environment.
- Improved sustainability. These compact systems and purification kits have reduced environmental impact vs. previous generation systems and cartridges. Polishing cartridges are 33% smaller and lighter, Lab Close mode saves water and energy, and packaging is greener. Plus, the new Drain Cap reduces the end-of-life impact of exhausted purification cartridges.
- Tailored media optimizes performance. Purification cartridges can be adapted to your feed water situation, whether extra hard tap water, or Elix® or non-Elix® pure water feed.
- Online monitoring & remote services. Securely manage your system from anywhere to support optimal productivity.
Compare Ultrapure Water System Features
Review the comparison table below to understand the similarities and differences between Milli-Q® IQ and EQ 7 series ultrapure water systems.
Features | Milli-Q® EQ 7000 | Milli-Q® EQ 7008/16 | Milli-Q® IQ 7000 | Milli-Q® IQ 7003/05/10/15 |
---|---|---|---|---|
Final water quality grade | Type 1 (ultrapure) | |||
Intermediate water quality | N/A | Type 3 (RO) | N/A | Type 2 (Elix®) |
Feed water quality | Pretreated* | Tap | Pretreated* | Tap |
Intermediate production flow rate (L/hr) | N/A | 8 or 16 | N/A | 3, 5, 10 or 15 |
IPAK Gard® pretreatment pack | No | Yes | No | Yes |
IPAK Meta® polishing cartridge | Yes | Yes | Yes | No |
IPAK Quanta® polishing cartridge | Yes | |||
Application POD-Paks | 6 | |||
Touchscreen interface | Yes | |||
MyMilli-Q™ Remote Care capable | Yes | |||
Lab Close mode | Yes | |||
Local network connectivity and USB port | Yes | |||
System footprint | ++++ | |||
Flexibility of installation | +++ | ++++ | ||
Flexibility of Q-POD® dispenser | +++ | ++++ | ||
Number of Q-POD® dispensers | 1 | Up to 4 | ||
Dispensing flow rates | 3 | 8 | ||
Volumetric dispensing | ++ | ++++ | ||
Drop-by-drop dispensing | No | Yes | ||
Assisted dispensing | No | Yes | ||
Data management | ++ | ++++ | ||
Dispense reports & QR codes | No | Yes | ||
Oxidative UV lamp | Mercury | Mercury-free [Design for Sustainability scorecard] | ||
TOC accuracy | In-line Indicator | A10® Monitor | ||
Compatible with Milli-Q® IQ Element unit (Brochure) | No | Yes | ||
Qualification | No | Yes | ||
TOC calibration/verification | No† | Yes | ||
USP suitability testing | No | Yes |
SELECT THE TYPE 1 WATER Purification SYSTEM THAT WORKS FOR YOU
The selection of the proper ultrapure water system for your laboratory will depend on several parameters, such as:
- Available feed water
- Daily volume needs
- Monitoring requirements
- Expected certification levels
Contact our Lab Water Specialists to discuss the ultrapure water system that’s best adapted to your needs.
Select and configure the optimal ultrapure water system for your lab.
Related Resources
Tech Article: Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Water System
Tech Article: Milli-Q® IQ 7000 Ultrapure Water System
Brochure: Milli-Q® EQ 7008/16 Ultrapure & Pure (RO) Water System
Brochure: Milli-Q® EQ 7000 Ultrapure Water System
Brochure: Milli-Q® IQ Element for Trace Elemental Analysis
Sustainability and Milli-Q® Lab Water Solutions
R&D Notebook: Bactericidal UVC LEDs for Ultrapure Water Applications
Tech Article: Drinking Water Testing by Ion Chromatography using Ultrapure Water
Tech Article: Water for Silica-Sensitive Applications
Tech Article: Determination of Toxic Elements in Environmental Analyses
Tech Article: Determination of Elemental Impurities in Pharmaceuticals
Tech Article: Trace LC-MS/MS Analysis of Hormones in Tap Water
Tech Article: Water for Next-Generation Sequencing
Tech Article: BPA-Free Water for Embryology Research
Water Storage Tanks
Water Purification Consumables
Application POD-Pak Final Filters