Nitrate Detection in Wastewater
Reflectometric determination after reduction to nitrite and reaction with Griess reagent.
Introduction
Excess nitrate in wastewater can lead to contamination of groundwater and subsequent environmental problems including eutrophication— excessive growth of algae and plants— upsetting marine ecosystems.1 In addition, human consumption of excess nitrate has been linked to cancer and a blood condition called methaemoglobinaemia.2 As a result, monitoring nitrate levels is critical for both environmental and health reasons. Here we describe nitrate determination in wastewater using reflectometry and the Reflectoquant® Nitrate Test Kit.
Experimental Method
Nitrate ions are reduced to nitrite ions by a reducing agent. In the presence of an acidic buffer, these nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts with N-(1-naphthyl)-ethylenediamine to form a red-violet azo dye that is determined reflectometrically.
Instruments
- Reflectometer RQflex® 20 Reflectoquant® (1.17246)*
*Note: This application note pertains to RQflex® 20 and all discontinued instruments (RQflex® 10, RQflex® plus).
Optional semi-quantitative determination
- Nitrate Test Method: colorimetric with test strips 10 - 25 - 50 - 100 - 250 - 500 mg/l NO₃⁻ MQuant® (1.10020)
Instrumental Analysis
- Press the reflectometer START key and simultaneously dip the test strip into the sample (15 - 30 °C) for ca. 2 seconds, ensuring that both reaction zones are immersed.
- Allow excess liquid to run off via the long edge of the strip on to an absorbent paper towel.
- Wait 60 seconds and then measure the strip in the reflectometer.
The value [mg/l] will be stored automatically. (Please refer to the RQflex® operating instructions and the instruction for use of the Reflectoquant® Nitrate Test.)
Note: If a semi-quantitative determination is sufficient there is the possibility to use MQuant® test strips, Cat. No. 1.10020 10 - 25 - 50 - 100 - 250 - 500 mg/l NO₃⁻
Procedure
Samples containing more than 500 mg/l NO3- must be diluted with distilled water. The pH must be within the range 1 - 12. If the pH is lower than 1, buffer the sample with sodium acetate; if it is greater than 12, adjust to approx. 3 - 5 with tartaric acid.
- Immerse both reaction zones of the test strip in the sample (15 - 25 °C) for 1 sec.
- Shake off excess liquid from the strip and after 1 min determine with which color field on the label the color of the NO3- reaction zone coincides most exactly.
- If the NO2- alert zone changes color, see “Notes on the measurement” in the instruction for use.
- Read off the corresponding result in mg/l NO3- or NO3-N.
Sample | Reflectoquant® [mg/l] | Photometry [mg/l] |
---|---|---|
1 | 21 | 19 |
2 | 56 | 54 |
3 | 47 | 38 |
4 | 14 | 14 |
5 | 12 | 10 |
6 | 15 | 14 |
7 | 20 | 18 |
8 | 18 | 17 |
9 | 14 | 15 |
10 | 18 | 18 |
11 | 12 | 12 |
12 | 20 | 21 |
13 | 18 | 21 |
14 | 64 | 66 |
15 | 37 | 40 |
16 | 6 | 7 |
17 | 3 | 3 |
18 | 19 | 16 |
19 | 24 | 21 |
20 | 28 | 30 |