Cell Culture Media Preparation Using MilliShot™ Single Dose Antibiotics
Why Use Antibiotics?
Antibiotics are crucial reagents for cell culture researchers to prevent contamination of cultures from pathogens and to minimize the loss of reagents and cells. Bacterial and fungal contaminants can induce physiological and morphological changes in cultured cells, affecting experimental outputs. Antibiotics can also be used to select for genetically modified cells during experimental assays. Therefore, antibiotics, in addition to aseptic conditions, are vital for reproducible tissue culture data.
While the most frequently used antibiotics to prevent contamination are Penicillin-Streptomycin or Antibiotic-Antimycotic solution, some scientists may add Amphotericin B or Gentamicin to the medium. Here we describe the protocol for preparing culture media using our MilliShot™ single dose antibiotics. MilliShot™ single dose antibiotic solutions are cell culture tested, ready-to-use antibiotics, conveniently packaged in one-time use vials, saving time spent preparing cell culture media.
Important Points to Note Before You Start
- All steps of media preparation and cell culture must be performed in an aseptic laminar flow hood.
- Confirm the right media and supplements for your cell line. Download the ECACC handbook for all information connected to the cell line of your interest.
- Bring your liquid media, FBS and supplements to 37°C before you start.
MilliShot™ Single Dose Antibiotics Protocol
Step 1
![Gloved hand removed a small vial from a plastic box that contains multiple vials. The box is from the freezer and has frost on it. Gloved hand removed a small vial from a plastic box that contains multiple vials. The box is from the freezer and has frost on it.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/protocols/cell-culture-and-analysis/media-preparation/removing-cells-from-freezer/removing-cells-from-freezer.jpg)
Step 2
![Full bottle of cell culture medium. Bottle has a label on the front that has the name of the medium, Dulbecco's Modified Eagle's Medium. Full bottle of cell culture medium. Bottle has a label on the front that has the name of the medium, Dulbecco's Modified Eagle's Medium.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/protocols/cell-culture-and-analysis/media-preparation/dmem-medium-bottle/dmem-medium-bottle.jpg)
Step 3
![Female scientist wearing a lab coat, gloves, and safety goggles sits at a tissue culture hood filtering cell culture media into another bottle. Female scientist wearing a lab coat, gloves, and safety goggles sits at a tissue culture hood filtering cell culture media into another bottle.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/protocols/cell-culture-and-analysis/media-preparation/sterile-filter/sterile-filter.jpg)
Step 4
![Scientist sits at the tissue culture hood with a six well tissue culture plate. In one hand he holds tweezers that hold onto a Millicell hanging insert, and in the other hand he holds a pipette that is changing the media in the insert. Scientist sits at the tissue culture hood with a six well tissue culture plate. In one hand he holds tweezers that hold onto a Millicell hanging insert, and in the other hand he holds a pipette that is changing the media in the insert.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/protocols/cell-culture-and-analysis/media-preparation/culturing-cells-millicell-inserts/culturing-cells-millicell-inserts.jpg)
All the steps are performed in laminar flow chamber under aseptic conditions.
- Open a 500mL bottle of medium suitable for your cell line, such as DMEM, RPMI, or MEM with 2mM glutamine.
- Add 50mL of FBS of your choice to prepare medium with 10% FBS. For cell lines that need 5% FBS supplementation, add 25mL of FBS of your choice.
- If your medium does not have added glutamine, add 5mL of 2mM glutamine. Add appropriate concentration of any other growth supplements that your cell line may require for growth.
- Open and add the entire contents (5mL) of one vial of MilliShot™ single dose Penicillin-Streptomycin/Antibiotic-Antimycotic solution/Amphotericin B/Gentamicin to the medium. You can directly pour or use a pipette to add the vial contents to the medium.
- Filter the prepared complete medium using Stericup® filtration devices to obtain sterile complete medium to culture your cell lines.
- Culture your cells as required for your cell type and experimental outcome.