MilliporeSigma
HomeCross-CouplingPEPPSI™-IPent for Demanding Cross-Coupling Reactions
PEPPSI™-IPent for Demanding Cross-Coupling Reactions
Introduction
Professor Mike Organ and co-workers have developed the PEPPSI™ (Pyridine-Enhanced Precatalyst Preparation Stabilization and Initiation) precatalysts for palladium-catalyzed cross-coupling reactions. PEPPSI™ palladium precatalysts have become a choice alternative to palladium phosphine complexes, as they are comprised of a bulky N-heterocyclic-carbene (NHC) ligand bonded to palladium and a s-donating 3-chloropyridine ligand for stability. The PEPPSI™-IPent (732117) further advances the agility of cross-coupling reactions by being air-stable and thus increasing user-friendliness. We are proud to offer the most active of the PEPPSI™ series, PEPPSI™-IPent, in collaboration with the Organ Research Group.1
peppsi-ipent
Advantages
  • PEPPSI™-IPent is bench stable: no need for a glovebox
  • Poisoning of Pd has not been observed
  • Higher pyrrole-based cross-coupling reactions success
  • Consistently superior coupling yields over PEPPSI™-IPr
  • Effective and versatile catalyst for difficult, sterically-demanding reactions2
peppsi ipent bench
Representative Applications
Under relatively mild conditions, PEPPSI™-IPent is a highly effective catalyst in challenging Suzuki–Miyaura and Negishi cross-coupling reactions. This produces tetra-ortho-substituted (hetero)biaryl compounds in high yields, especially when compared to the use of PEPPSI™-IPr. Bulky and acidic substituents were observed to be well tolerated.1,2
peppsi ipent catalyst
Negishi cross-coupling reactions of secondary alkyl zinc halides with aryl halides utilize PEPPSI™-IPent, boasting higher selectivity than PEPPSI™-IPr. The desired branched product was obtained in higher yields than that of the linear product.1,3
peppsi ipent ipr
Buchwald–Hartwig–Yagupol’skii aryl aminations typically use strong bases to aid the rate-limiting step, creating conditions that functional groups poorly tolerate. In contrast, using mildly basic conditions and PEPPSI™-IPent, a variety of demanding aryl chlorides were successfully coupled to secondary amines. In addition, PEPPSI™-IPent facilitated the coupling of electron-deficient anilines with electron-rich aryl chlorides. In these reactions, the highly reactive PEPPSI™-IPent consistently outperformed PEPPSI™-IPr.1,4,5
peppsi ipent toluene
In Stille–Migita cross-couplings of heteroaryl stannanes with aryl halides, PEPPSI™-IPent allowed for lower reaction temperatures below 100 ºC for thermally sensitive cross-coupling partners. Organ and co-workers observed successful coupling within in the range of 30–80 ºC while maintaining consistent yields.1,6
peppsi ipent stille
High temperatures frequently used for carbon-sulfur bond formation were circumvented with PEPPSI™-IPent, featuring mild conditions for the formation of thiol ethers by metal-catalyzed sulfinations. Successful reactions have included the sulfination of (hetero)aryl halides with aryl sulfides, alkyl thiols, and also sterically hindered substrates.1,7
peppsi ipent temperature
Materials
Loading
References
1.
Valente C, Çalimsiz S, Hoi KH, Mallik D, Sayah M, Organ MG. 2012. The Development of Bulky Palladium NHC Complexes for the Most-Challenging Cross-Coupling Reactions. Angew. Chem. Int. Ed.. 51(14):3314-3332. https://doi.org/10.1002/anie.201106131
2.
Organ MG, Calimisiz S, Sayah M, Hoi K, Lough A. 2009. Angew. Chem. Int. Ed.. 482283–7.
3.
Çalimsiz S, Organ MG. 2011. Negishi cross-coupling of secondary alkylzinc halides with aryl/heteroaryl halides using Pd?PEPPSI?IPent. Chem. Commun.. 47(18):5181. https://doi.org/10.1039/c0cc04835f
4.
Hoi KH, Çalimsiz S, Froese RDJ, Hopkinson AC, Organ MG. 2011. Amination with Pd?NHC Complexes: Rate and Computational Studies on the Effects of the Oxidative Addition Partner. Chem. Eur. J.. 17(11):3086-3090. https://doi.org/10.1002/chem.201002988
5.
Hoi KH, Çalimsiz S, Froese RDJ, Hopkinson AC, Organ MG. 2012. Amination with Pd?NHC Complexes: Rate and Computational Studies Involving Substituted Aniline Substrates. Chem. Eur. J.. 18(1):145-151. https://doi.org/10.1002/chem.201102428
6.
Dowlut M, Mallik D, Organ M. 2010. An Efficient Low-Temperature Stille-Migita Cross-Coupling Reaction for Heteroaromatic Compounds by Pd-PEPPSI-IPent. Chem. Eur. J.. 16(14):4279-4283. https://doi.org/10.1002/chem.200903337
7.
Sayah M, Organ MG. 2011. Carbon-Sulfur Bond Formation of Challenging Substrates at Low Temperature by Using Pd-PEPPSI-IPent. Chem. Eur. J.. 17(42):11719-11722. https://doi.org/10.1002/chem.201102158