Skip to Content
Merck
All Photos(1)

Key Documents

APT129

Sigma-Aldrich

Caspase 8 Colorimetric Activity Assay Kit 25, IETD

Caspase-8 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the sequence IETD.

Synonym(s):

FLICE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

UniProt accession no.

detection method

colorimetric

shipped in

dry ice

General description

Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. Chemicon′s Caspase-8 Colorimetric Activity Assay Kits provide a simple and convenient means for assaying the activity of caspases that recognize the sequence IETD. The assay is based on spectophotometric detection of the chromophore p-nitroaniline (pNA) after cleavage from the labeled substrate IETD-pNA. The free pNA can be quantified using a spectrophotometer or a microtiter plate reader at 405 nm. Comparison of the absorbance of pNA from an apoptotic sample with an uninduced control allows determination of the fold increase in caspase-8 activity.

Application

Research Category
Apoptosis & Cancer

Components

5X Cell Lysis Buffer (Part No. 90065): 5 mL

5X Assay Buffer (Part No. 90066): 10 mL

Caspase-8 Substrate (Ac-IETD-pNA) (Part No. 90081): 250 μL of 2.5 mg/mL solution

Caspase-8 Inhibitor (Ac-IETD-CHO) (Part No. 90086): 50 μL of 100 mM (0.05 mg/mL) in DMSO

pNA Standard (Part No. 90085): 250 μL of 10 mM in DMSO

Storage and Stability

Store kit materials at -20°C up to their expiration date.

Special Precautions:

· After thawing reagents, use immediately or aliquot and freeze at -20oC for longer storage. Avoid repeated freeze/thaw cycles.

· The Caspase-8 substrate and pNA standard are especially light sensitive. Maintain these reagents in amber or covered containers.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raul Chavez-Valdez et al.
Frontiers in physiology, 10, 306-306 (2019-04-12)
Toll-like receptor (TLR)3 activation during the neonatal period produces responses linked to the origins of neuropsychiatric disorders. Although there is sexual dimorphism in neuropsychiatric disorders, it is unknown if brain responses to TLR3 activation are sex-specific. We hypothesized that poly
Udeshika Yapa Bandara et al.
Asian Pacific journal of cancer prevention : APJCP, 23(2), 501-510 (2022-03-01)
Pomegranate ,a polyphenol-rich fruit, has been considered as one of the ancient fruits with anticancer effect. Cell cycle arrest is considered as an ordinary factor in human cancer, and apoptosis is the frequent drug target. This study aimed to evaluate
Christopher J Anderson et al.
Nature, 596(7871), 262-267 (2021-08-06)
Regulated cell death is an integral part of life, and has broad effects on organism development and homeostasis1. Malfunctions within the regulated cell death process, including the clearance of dying cells, can manifest in diverse pathologies throughout various tissues including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service