Merck
Z512974
Sigma-Aldrich
Aldrich® 腐食性レクチャーボトルガスレギュレーター
delivery pressure 0-50 psig, CGA Inlet size 170
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41100000
NACRES:
NB.51
delivery pressure measuring range
0-50 psig
CGA注入口サイズ
170
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
  • Single-stage
  • CGA inlet connection, needle valve outlet with ¼ in. NPTM connection
  • Max. inlet pressure: 0-4000 psig
  • Temp. range: -40 to 140°F
  • 316 Stainless Steel body 1 ½ in. gauge face, PTFE seals and inner friction sleeves
  • NPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure; order bonnet-vent tubing connectors separately
  • Use with "T"-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipment
  • Use for high-purity applications
Lecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
その他情報
テクニカルインフォメーションAL-151 Gas Regulators: Selection, Installation and Operationをご覧ください。
法的情報
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
最新バージョンのいずれかを選択してください:
試験成績書(COA)