コンテンツへスキップ
Merck
すべての画像(1)

主要文書

安全性情報

MABF567

Sigma-Aldrich

Anti-CD4 Antibody (mouse), violetFluor® 450, clone GK1.5

clone GK1.5, 0.2 mg/mL, violetFluor®

別名:

T-cell surface glycoprotein CD4, T-cell differentiation antigen L3T4, T-cell surface antigen T4/Leu-3, CD4

ログイン組織・契約価格を表示する


About This Item

UNSPSCコード:
12352203
eCl@ss:
32160702
NACRES:
NA.41
結合体:
violetFluor® 450
application:
FACS
IHC
IP
クローン:
GK1.5, monoclonal
化学種の反応性:
mouse
citations:
テクニック:
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

由来生物

rat

結合体

violetFluor® 450

抗体製品の状態

purified antibody

抗体製品タイプ

primary antibodies

クローン

GK1.5, monoclonal

化学種の反応性

mouse

包装

antibody small pack of 25 μg

メーカー/製品名

violetFluor®

濃度

0.2 mg/mL

テクニック

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

アイソタイプ

IgG2bκ

UniProtアクセッション番号

輸送温度

wet ice

ターゲットの翻訳後修飾

unmodified

遺伝子情報

mouse ... Cd4(12504)

詳細

The GK1.5 antibody reacts with mouse CD4, a 55 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The GK1.5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the GK1.5 antibody and an alternative antibody, Anti-Mouse CD4 clone RM4-5, will “compete” for binding, i.e. RM4-5 is able to block GK1.5 binding to cells. In contrast, the Anti-Mouse CD4 clone RM4-4 does not block binding of the GK1.5 antibody to cells (Arora S et al. 2006. Infect. Immun. 74: 4339-4348). The GK1.5 antibody is also reported to be cross-reactive with Syrian hamster CD4.

免疫原

Immunogen information for this antibody is proprietary information.

アプリケーション

Research Category
炎症及び免疫
This rat monoclonal antibody Anti-CD4 (mouse), violetFluor 450, clone GK1.5 validated for use in IP, IHC & Flow Cytometry for the detection of CD4.

品質

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.25 µg of this antibody detected CD4 in one million C57Bl/6 splenocytes.

物理的形状

Size Exclusion Chromatography
Purified Rat Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.

保管および安定性

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

法的情報

violetFluor is a registered trademark of Tonbo Biotechnologies

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

適切な製品が見つかりませんか。  

製品選択ツール.をお試しください

保管分類コード

12 - Non Combustible Liquids

WGK

nwg

引火点(°F)

Not applicable

引火点(℃)

Not applicable


適用法令

試験研究用途を考慮した関連法令を主に挙げております。化学物質以外については、一部の情報のみ提供しています。 製品を安全かつ合法的に使用することは、使用者の義務です。最新情報により修正される場合があります。WEBの反映には時間を要することがあるため、適宜SDSをご参照ください。

Jan Code

MABF567:


試験成績書(COA)

製品のロット番号・バッチ番号を入力して、試験成績書(COA) を検索できます。ロット番号・バッチ番号は、製品ラベルに「Lot」または「Batch」に続いて記載されています。

以前この製品を購入いただいたことがある場合

文書ライブラリで、最近購入した製品の文書を検索できます。

文書ライブラリにアクセスする

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

製品に関するお問い合わせはこちら(テクニカルサービス)