์ฝ˜ํ…์ธ ๋กœ ๊ฑด๋„ˆ๋›ฐ๊ธฐ
Merck
๋ชจ๋“  ์‚ฌ์ง„(1)
Key Documents
Z105813
Polyethylene stopper
joint: ST/NS 14/20
๋™์˜์–ด(๋“ค):
flask stopper, flask stoppers, stoppers
๋กœ๊ทธ์ธ์กฐ์ง ๋ฐ ๊ณ„์•ฝ ๊ฐ€๊ฒฉ ๋ณด๊ธฐ
About This Item
UNSPSC ์ฝ”๋“œ:
41122500
NACRES:
NB.35
์กฐ์ธํŠธ
joint: ST/NS14/20
์œ ์‚ฌํ•œ ์ œํ’ˆ์„ ์ฐพ์œผ์‹ญ๋‹ˆ๊นŒ? ๋ฐฉ๋ฌธ ์ œํ’ˆ ๋น„๊ต ์•ˆ๋‚ด
์ผ๋ฐ˜ ์„ค๋ช…
Nonthreaded, wide, thick flanges.
์‹œํ—˜ ์„ฑ์ ์„œ(COA)
์ œํ’ˆ์˜ ๋กœํŠธ/๋ฐฐ์น˜ ๋ฒˆํ˜ธ๋ฅผ ์ž…๋ ฅํ•˜์—ฌ ์‹œํ—˜ ์„ฑ์ ์„œ(COA)์„ ๊ฒ€์ƒ‰ํ•˜์‹ญ์‹œ์˜ค. ๋กœํŠธ ๋ฐ ๋ฐฐ์น˜ ๋ฒˆํ˜ธ๋Š” ์ œํ’ˆ ๋ผ๋ฒจ์— ์žˆ๋Š” โ€˜๋กœํŠธโ€™ ๋˜๋Š” โ€˜๋ฐฐ์น˜โ€™๋ผ๋Š” ์šฉ์–ด ๋’ค์—์„œ ์ฐพ์„ ์ˆ˜ ์žˆ์Šต๋‹ˆ๋‹ค.
์ด ์ œํ’ˆ์„ ์ด๋ฏธ ๊ฐ€์ง€๊ณ  ๊ณ„์‹ญ๋‹ˆ๊นŒ?
๋ฌธ์„œ ๋ผ์ด๋ธŒ๋Ÿฌ๋ฆฌ์—์„œ ์ตœ๊ทผ์— ๊ตฌ๋งคํ•œ ์ œํ’ˆ์— ๋Œ€ํ•œ ๋ฌธ์„œ๋ฅผ ์ฐพ์•„๋ณด์„ธ์š”.
๋ฌธ์„œ ๋ผ์ด๋ธŒ๋Ÿฌ๋ฆฌ ๋ฐฉ๋ฌธ
์ด๋ฏธ ์—ด๋žŒํ•œ ๊ณ ๊ฐ
Slide 1 of 7
1 of 7
Precision Seal® rubber septa white, 24/40 joints
Sigma-Aldrich
Z553980
Precision Sealยฎ rubber septa
Precision Seal® rubber septa white, 14/20 joints
Sigma-Aldrich
Z553964
Precision Sealยฎ rubber septa
10-Undecen-1-ol 98%
Sigma-Aldrich
U2008
10-Undecen-1-ol
Calcium hydride powder, 0-2 mm, reagent grade, ≥90% (gas-volumetric)
Sigma-Aldrich
213268
Calcium hydride
Precision Seal® rubber septa white, 19/22 joints
Sigma-Aldrich
Z553972
Precision Sealยฎ rubber septa
Tetrahydrothiophene 99%
Sigma-Aldrich
T15601
Tetrahydrothiophene
์ž์‚ฌ์˜ ๊ณผํ•™์žํŒ€์€ ์ƒ๋ช… ๊ณผํ•™, ์žฌ๋ฃŒ ๊ณผํ•™, ํ™”ํ•™ ํ•ฉ์„ฑ, ํฌ๋กœ๋งˆํ† ๊ทธ๋ž˜ํ”ผ, ๋ถ„์„ ๋ฐ ๊ธฐํƒ€ ๋งŽ์€ ์˜์—ญ์„ ํฌํ•จํ•œ ๋ชจ๋“  ๊ณผํ•™ ๋ถ„์•ผ์— ๊ฒฝํ—˜์ด ์žˆ์Šต๋‹ˆ๋‹ค..
๊ณ ๊ฐ์ง€์›ํŒ€์œผ๋กœ ์—ฐ๋ฝ๋ฐ”๋ž๋‹ˆ๋‹ค.