Skip to Content
Merck
All Photos(1)

Key Documents

28943

Millipore

Andrade peptone water, Vegitone

suitable for microbiology

Synonym(s):

Peptone water Andrade, Vegitone, Vegitone Andrade peptone water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
clinical testing
food and beverages

microbiology

suitability

Providencia spp.
nonselective and differential for Citrobacter spp.
nonselective and differential for Enterobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for Yersinia spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

Andrade Peptone Water is a basal medium to which various carbohydrates can be added to study fermentation reactions, particularly of members of the Enterobacteriaceae. Media from the Vegitone-line do not contain animal derived material[1]

Components

Components (g/L):
andrade inicator 0.1
Peptone (vegetable) 10.0
sodium chloride 5.0

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.I. Barrow, R.K.A. Feltham
Cowan and Steel?s Manual for the Identification of Medical Bacteria, 161-163 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service