Recommended Products
shelf life
limited shelf life, expiry date on the label
Quality Level
composition
ammonium ferric citrate, 0.5 g/L
bromocresol purple, 0.02 g/L
D(+)-glucose, 1 g/L
Ingredients: agar, 12.5 g/L
L-lysine monohydrochloride, 10 g/L
meat peptone, 5 g/L
sodium thiosulfate, 0.04 g/L
yeast extract, 3 g/L
final pH
6.7±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
nonselective and differential for Citrobacter spp.
nonselective and differential for Enterococcus spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service