Skip to Content
Merck
All Photos(2)

Key Documents

OGS268

Sigma-Aldrich

PSF-AD5 - ADENOVIRUS TYPE 5 PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

form

buffered aqueous solution

mol wt

size 4034 bp

bacteria selection

kanamycin

origin of replication

BR322 (15 copies)

peptide cleavage

no cleavage

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This is a serotype 5 adenovirus virus vector that allows the insertion of genes into the E1A position within the virus. The vector contains an E1 and E3 deletion and has been modified to remove a number of key resitrction sites. The plasmid contains a small multiple cloning site consisting of AsiSI ClaI and PacI. These sites will be found in all of the plasmid vectors in our product range which allows the direct cloning of segments from our catalogue plasmids into the adenovirus genome (see cloning tab for more details). The vector also contains a blue white screening cassette that allows for the detection of bacterial clones carrying any newly inserted DNA.

Packaging Capacity: The wiltype virus genome is 35938 base pairs but in this vector the genome has been shortended by the deletions to make it 30501 base pairs. This allows for the insertion of up to 5. 5kb of exogenous DNA. It has also previously been shown that serotype 5 adenovirus genomes can accomdate insertions up to 105% total genome length compared to wild type. For this reason it should be possible to insert an additional 1. 7kb resulting in a total insertion size of 7. 2Kb. We have previously packaged viruses up to 105% of the virus genome with good stability but some insertions may be unstable at this length depending on sequence. These numbers are therefore provided as guidelines only.

Recovery and Growth: Virus recovery requires two steps firstly the plasmid must be cleaved with the SwaI restriction enzyme to linearise the virus genome. Then the linear genome must be recovered and grown in 293 cells or any derivative of 293 cells such as 293As and 293T cells. These lines contain and express the adenovirus E1A protein that transcompliments the deletion in the E1A region in this plasmid. A typically removery transfection would use approximately 5ug of DNA. It is not neccessary to remove the plasmid backbone after linearising the DNA a standard DNA clean up will be sufficient rather than gel extraction. Virus colonies should appear approximately 10-15 days after transfection into 293 cells; however, clones can sometimes appear at earlier time points. Cell should be cultured in 2% fetal calf serum to prevent over growth during this time.

Promoter Expression Level:

Application

This plasmid contains a serotype 5 Adenovirus non-replicating vector that allows for the insertion of expression cassettes into the E1A region (5 prime end of the genome). The vector can be grown in 293 cells (which express Adenovirus E1A) to produce high titre adenovirus preparations.

To clone in an expression cassette into this virus genome there are multiple possible strategies. You can either directly clone any expression cassette from our product range by cutting the plasmid with AsiSI and PacI and cloning the cassette into the same sites in the virus genome. You can also clone this fragment into just the AsiSI site or the PacI site because these sites have the same overhang. We recommend cloning into just the PacI site where possible because the efficiency of cutting with AsiSI is typically not 100%.

Alternatively you can use Gibson Assembly InFusionHD Seamless GeneArt or LIC to clone in the expression cassette. This can be achieved by amplifying the expression cassette from any of our plasmids using these primers:
Forward primer if cutting the virus genome with AsiSI:
GAAACAAATTCTAGGAGGTACTCCGTATGGCGATCGCGGCTCCCGACATC
Forward primer if cutting the virus genome with PacI:
TGGGGCGGCCGCCATCGATGGGTTAATTAAGGCTCCCGACATCTTGG
Reverse primer if cutting the virus genome with PacI or PacI and AsiSI:
GGCACTTCCAGCAGCAGTTTCGGATCGCCGTTAATTAAGTTTCGATAGCC
These primers will bind to almost our entire product range of plasmids with the forward primers binding upstream of the promoter (flanked by Bgl2 in our vectors) and the reverse primer binding downstream of the terminator after the MCS. This allows the entire promoter-gene-terminator section to be amplified. The primers also contain arms of homology to the MCS in the virus genome which allows the PCR amplified fragment to be inserted using any of the techniques named above.

Sequence

To view sequence information for this product, please visit the product page

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.

Articles

SnapFast™ plasmid system eliminates restriction sites in DNA sections, ensuring flexibility and functionality in molecular cloning..

Versatile sequencing primers enable sequencing of inserts in plasmids at specific positions, aiding in molecular biology research.

Plasmid platform with interchangeable DNA components offers versatile research tools for genetic studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service