Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47885-U

Supelco

Supelco® 37 Component FAME Mix

10 mg/mL in methylene chloride (varied), analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

EC Number:
UNSPSC Code:
85151701

grade

analytical standard

packaging

ampule of 1 mL

concentration

10 mg/mL in methylene chloride (varied)

shipped in

dry ice

storage temp.

−20°C

General description

This product has been upgraded to a certified reference material (CRM). Please order using catalog number CRM47885.

Use this qualitative standard to identify key fatty acid methyl esters in a variety of food products. The complex nature of this mix eliminates the need to purchase multiple mixtures for identification while minimizing expenses.

For information on the chromatographic analysis of this standard, please contact Technical Services at techservice@sial.com.

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    cis-13,16-Docosadienoic acid methyl ester 2 wt. %
    cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 2 wt. %
    cis-11,14-Eicosadienoic acid methyl ester 2 wt. %
    cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester 2 wt. %
    cis-8,11,14-Eicosatrienoic acid methyl ester 2 wt. %
    cis-11,14,17-Eicosatrienoic acid methyl ester 2 wt. %
    cis-11-Eicosenoic acid methyl ester 2 wt. %
    Methyl cis-10-heptadecenoate 2 wt. %
    Methyl hexanoate 4 wt. %
    Methyl γ-linolenate 2 wt. %
    Methyl arachidate 4 wt. %
    Methyl arachidonate 2 wt. %
    Methyl behenate 4 wt. %
    Methyl butyrate 4 wt. %
    Methyl decanoate 4 wt. %
    Methyl dodecanoate 4 wt. %
    Methyl elaidate 2 wt. %
    Methyl erucate 2 wt. %
    Methyl heneicosanoate 2 wt. %
    Methyl heptadecanoate 2 wt. %
    Methyl linoleate 2 wt. %
    Methyl linolelaidate 2 wt. %
    Methyl linolenate 2 wt. %
    Methyl myristate 4 wt. %
    Methyl myristoleate 2 wt. %
    Methyl oleate 4 wt. %
    Methyl octanoate 4 wt. %
    Methyl palmitate 6 wt. %
    Methyl palmitoleate 2 wt. %
    Methyl pentadecanoate 2 wt. %
    Methyl cis-10-pentadecenoate 2 wt. %
    Methyl stearate 4 wt. %
    Methyl tricosanoate 2 wt. %
    Methyl tetracosanoate 4 wt. %
    Methyl tridecanoate 2 wt. %
    Methyl undecanoate 2 wt. %
    Methyl cis-15-tetracosenoate 2 wt. %
    See All (37)

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P Bougnoux et al.
British journal of cancer, 101(12), 1978-1985 (2009-11-19)
Breast cancer becomes lethal when visceral metastases develop. At this stage, anti-cancer treatments aim at relieving symptoms and delaying death without resulting in additional toxicity. On the basis of their differential anti-oxidant defence level, tumour cells can be made more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service