MilliporeSigma
Human NLRP1 is a sensor for double-stranded RNA.
Science (New York, N.Y.) (2020-11-28)
Stefan Bauernfried, Matthias J Scherr, Andreas Pichlmair, Karl E Duderstadt, Veit Hornung
ABSTRACT
Inflammasomes function as intracellular sensors of pathogen infection or cellular perturbation and thereby play a central role in numerous diseases. Given the high abundance of NLRP1 in epithelial barrier tissues, we screened a diverse panel of viruses for inflammasome activation in keratinocytes. We identified Semliki Forest virus (SFV), a positive-strand RNA virus, as a potent activator of human but not murine NLRP1B. SFV replication and the associated formation of double-stranded (ds) RNA was required to engage the NLRP1 inflammasome. Moreover, delivery of long dsRNA was sufficient to trigger activation. Biochemical studies revealed that NLRP1 binds dsRNA through its leucine-rich repeat domain, resulting in its NACHT domain gaining adenosine triphosphatase activity. Altogether, these results establish human NLRP1 as a direct sensor for dsRNA and thus RNA virus infection.
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
TERGITOL solution, Type NP-40, 70% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Benzamidine, ≥95.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Ribavirin, antiviral
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Nigericin sodium salt, ≥98% (TLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Human Serum, Heat Inactivated (from male AB clotted whole blood), USA origin, sterile-filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Triton X-100, laboratory grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
$286.00
10 mL
In Stock
Details...
$475.00
$332.50