MilliporeSigma
  • Chemical modification of reactive multilayered films fabricated from poly(2-alkenyl azlactone)s: design of surfaces that prevent or promote mammalian cell adhesion and bacterial biofilm growth.
Chemical modification of reactive multilayered films fabricated from poly(2-alkenyl azlactone)s: design of surfaces that prevent or promote mammalian cell adhesion and bacterial biofilm growth.
Biomacromolecules (2009-05-15)
Maren E Buck, Anthony S Breitbach, Sonja K Belgrade, Helen E Blackwell, David M Lynn
ABSTRACT
We report an approach to the design of reactive polymer films that can be functionalized post-fabrication to either prevent or promote the attachment and growth of cells. Our approach is based on the reactive layer-by-layer assembly of covalently crosslinked thin films using a synthetic polyamine and a polymer containing reactive azlactone functionality. Our results demonstrate (i) that the residual azlactone functionality in these films can be exploited to immobilize amine-functionalized chemical motifs similar to those that promote or prevent cell and protein adhesion when assembled as self-assembled monolayers on gold-coated surfaces and (ii) that the immobilization of these motifs changes significantly the behaviors and interactions of cells with the surfaces of these polymer films. We demonstrate that films treated with the hydrophobic molecule decylamine support the attachment and growth of mammalian cells in vitro. In contrast, films treated with the hydrophilic carbohydrate d-glucamine prevent cell adhesion and growth almost completely. The results of additional experiments suggest that these large differences in cell behavior can be understood, at least in part, in terms of differences in the abilities of these two different chemical motifs to promote or prevent the adsorption of protein onto film-coated surfaces. We demonstrate further that this approach can be used to pattern regions of these reactive films that resist the initial attachment and subsequent invasion of mammalian cells for periods of at least one month in the presence of serum-containing cell culture media. Finally, we report that films that prevent the adhesion and growth of mammalian cells also prevent the initial formation of bacterial biofilms when incubated in the presence of the clinically relevant pathogen Pseudomonas aeruginosa . The results of these studies, collectively, suggest the basis of general approaches to the fabrication and functionalization of thin films that prevent, promote, or pattern cell growth or the formation of biofilms on surfaces of interest in the contexts of both fundamental biological studies and a broad range of other practical applications.
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, natural, ≥97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Supelco
Acetone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, viscous liquid, Low-peroxide
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, for molecular biology, syrup
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, viscous liquid, Preservative Free, Low-peroxide; Low-carbonyls
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mg
In Stock
Details...
$259.00