MilliporeSigma
  • Enzymatic activation of cellulose acetate membrane for reducing of protein fouling.
Enzymatic activation of cellulose acetate membrane for reducing of protein fouling.
Colloids and surfaces. B, Biointerfaces (2012-01-06)
Derya Y Koseoglu-Imer, Nadir Dizge, Ismail Koyuncu
ABSTRACT
In this study, the surface of cellulose acetate (CA) ultrafiltration membrane was activated with serine protease (Savinase) enzyme to reduce protein fouling. Enzyme molecules were covalently immobilized with glutaraldehyde (cross-linking agent) onto the surface of CA membranes. The membrane activation was verified using filtration experiments and morphological analysis. Scanning electron microscopy (SEM) images and attenuated total reflectance Fourier transform infrared (ATR-FTIR) spectroscopy of the activated membrane when compared with raw membrane were confirmed that the enzyme was immobilized onto the membrane surface. The immobilization efficiencies changed from 13.2 to 41.2% according to the enzyme ratios from 2.5 to 10.0 mg/mL. However, the permeability values decreased from 232±6 to 121±4 L/m(2) h bar with increasing enzyme concentration from 2.5 to 10.0 mg/mL. In fouling experiments, bovine serum albumin (BSA) was used as the protein model solution and activated sludge was used as the model biological sludge. Enzyme-activated membranes exhibited good filtration performances and protein rejection efficiencies were compared with raw CA membrane. Also the relative flux reduction (RFR) ratios of membranes were calculated as 97% and 88% for raw CA and enzyme-activated membranes (5 mg/mL savinase), respectively. The membrane activated with Savinase enzyme could be proposed as a surface treatment method before filtration to mitigate protein fouling.
Sigma-Aldrich
Proteinase, bacterial, Type XXIV, 7.0-14.0 units/mg solid, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Protease from Bacillus licheniformis, lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Protease from Bacillus licheniformis, ≥2.4 U/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00
Sigma-Aldrich
Protease from Bacillus licheniformis, glycerol solution (50%)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
Estimated to ship on March 22, 2025
$196.00
4 x 25 μL
Estimated to ship on March 22, 2025
$404.00