MilliporeSigma
  • Microneme protein 5 regulates the activity of Toxoplasma subtilisin 1 by mimicking a subtilisin prodomain.
Microneme protein 5 regulates the activity of Toxoplasma subtilisin 1 by mimicking a subtilisin prodomain.
The Journal of biological chemistry (2012-08-17)
Savvas Saouros, Zhicheng Dou, Maud Henry, Jan Marchant, Vern B Carruthers, Stephen Matthews
ABSTRACT
Toxoplasma gondii is the model parasite of the phylum Apicomplexa, which contains obligate intracellular parasites of medical and veterinary importance. Apicomplexans invade host cells by a multistep process involving the secretion of adhesive microneme protein (MIC) complexes. The subtilisin protease TgSUB1 trims several MICs on the parasite surface to activate gliding motility and host invasion. Although a previous study showed that expression of the secretory protein TgMIC5 suppresses TgSUB1 activity, the mechanism was unknown. Here, we solve the three-dimensional structure of TgMIC5 by nuclear magnetic resonance (NMR), revealing that it mimics a subtilisin prodomain including a flexible C-terminal peptide that may insert into the subtilisin active site. We show that TgMIC5 is an almost 50-fold more potent inhibitor of TgSUB1 activity than the small molecule inhibitor N-[N-(N-acetyl-L-leucyl)-L-leucyl]-L-norleucine (ALLN). Moreover, we demonstrate that TgMIC5 is retained on the parasite plasma membrane via its physical interaction with the membrane-anchored TgSUB1.
Sigma-Aldrich
Proteinase, bacterial, Type XXIV, 7.0-14.0 units/mg solid, lyophilized powder
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Protease from Bacillus licheniformis, lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Protease from Bacillus licheniformis, ≥2.4 U/g
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Protease from Bacillus licheniformis, glycerol solution (50%)
Pricing and Inventory is not available. Please check the Millipore website for more information.