MilliporeSigma
  • Cross talk between the nuclease and helicase activities of Dna2: role of an essential iron-sulfur cluster domain.
Cross talk between the nuclease and helicase activities of Dna2: role of an essential iron-sulfur cluster domain.
Nucleic acids research (2012-06-12)
Subhash Pokharel, Judith L Campbell
ABSTRACT
Dna2 nuclease/helicase is a multitasking protein involved in DNA replication and recombinational repair, and it is important for preservation of genomic stability. Yeast Dna2 protein contains a conserved putative Fe-S (iron-sulfur) cluster signature motif spanning the nuclease active site. We show that this motif is indeed an Fe-S cluster domain. Mutation of cysteines involved in metal coordination greatly reduces not just the nuclease activity but also the ATPase activity of Dna2, suggesting that the nuclease and helicase activities are coupled. The affinity for DNA is not significantly reduced, but binding mode in the C to A mutants is altered. Remarkably, a point mutation (P504S), proximal to the Fe-S cluster domain, which renders cells temperature sensitive, closely mimics the global defects of the Fe-S cluster mutation itself. This points to an important role of this conserved proline residue in stabilizing the Fe-S cluster. The C to A mutants are deficient in DNA replication and repair in vivo, and, strikingly, the degree to which they are defective correlates directly with degree of loss of enzymatic activity. Taken together with previous results showing that mutations in the ATP domain affect nuclease function, our results provide a new mechanistic paradigm for coupling between nuclease and helicase modules fused in the same polypeptide.
Sigma-Aldrich
Proteinase, bacterial, Type XXIV, 7.0-14.0 units/mg solid, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Protease from Bacillus licheniformis, lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Protease from Bacillus licheniformis, ≥2.4 U/g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Protease from Bacillus licheniformis, glycerol solution (50%)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00