MilliporeSigma
  • Novel coupling of surfactant assisted emulsification dispersive liquid-liquid microextraction with spectrophotometric determination for ultra trace nickel.
Novel coupling of surfactant assisted emulsification dispersive liquid-liquid microextraction with spectrophotometric determination for ultra trace nickel.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-26)
Qingwen Deng, Meihui Chen, Lamei Kong, Xia Zhao, Jie Guo, Xiaodong Wen
ABSTRACT
In this work, dispersive liquid-liquid microextraction (DLLME) was improved and the preconcentration method named as surfactant assisted emulsification dispersive liquid-liquid microextraction (SAE-DLLME) was established for ultra trace nickel preconcentration and spectrophotometric determination. Non-ionic surfactant Triton X-100 (TX-100) was used as emulsifier and Triton X-114 (TX-114) was investigated as comparison. Disperser solvent was substituted by surfactant, which could afford more effective emulsification and make the extraction relatively greener. The extraction was accomplished efficiently in only 1 min during manual shaking. Compared to traditional DLLME, the developed SAE-DLLME pretreatment was simple, rapid and effective. The improved extraction technique was firstly coupled with traditional spectrophotometer to improve the analytical performance and expand the application of spectrophotometric determination. The influence factors relevant to SAE-DLLME including extraction parameters and instrumental conditions, were studied in detail. Under the optimal conditions, the limit of detection (LOD) for nickel was 0.24 μg L(-1), with sensitivity enhancement factor (EF) of 23.
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, for molecular biology
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Triton X-114, laboratory grade
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard