MilliporeSigma
  • Layer-by-layer assembly of polyelectrolyte and graphene oxide for open-tubular capillary electrochromatography.
Layer-by-layer assembly of polyelectrolyte and graphene oxide for open-tubular capillary electrochromatography.
Journal of chromatography. A (2013-02-19)
Qishu Qu, Chenhao Gu, Zuli Gu, Yuqi Shen, Chengyin Wang, Xiaoya Hu
ABSTRACT
In this paper, open-tubular capillary column coated with graphene oxide (GO) was prepared through ionic adsorption of negatively charged GO nanosheets onto the capillary wall pre-modified with positively charged poly(diallydimethylammonium chloride) (PDDA). Thus prepared coating was very stable and could endure over 200 separations. The electroosmotic flow (EOF) characteristics of bare fused silica capillary column, PDDA coated column, and GO-PDDA coated column (GO-PDDA@column) were investigated by varying the percentage of methanol in buffer and the buffer pH value. The run-to-run, day-to-day, and column-to-column reproducibilities of EOF on GO-PDDA@column were satisfying with relative standard deviation values of less than 2% in all cases. The stationary phase displays a characteristic reversed-phase behavior. The GO-PDDA@column was also used to separate proteins in egg white. Both basic and acidic proteins were separated in a single run.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%