MilliporeSigma
  • The measurement of 1,2-propanediol, D, L-2,3-butanediol and meso-2,3-butanediol in controls and alcoholic cirrhotics.
The measurement of 1,2-propanediol, D, L-2,3-butanediol and meso-2,3-butanediol in controls and alcoholic cirrhotics.
Alcohol and alcoholism (Oxford, Oxfordshire). Supplement (1987-01-01)
J P Casazza, J Frietas, D Stambuk, M Y Morgan, R L Veech
ABSTRACT
Plasma 1,2-propanediol, D,L,-2,3-butanediol and 2,3-meso-butanediol were measured in 29 patients with biopsy proven alcoholic cirrhosis and 10 control subjects, none of whom had measurable blood alcohol levels. None of these compounds were present in control subjects at level above 5 nmol/ml. Seventeen (59%) of 29 serum samples drawn from patients with alcoholic cirrhosis contained 1,2-propanediol in concentrations above 5 nmol/ml (range less than 5.0 nmol/ml-35.4 nmol/ml, median 5.6), 8 (28%) of 29 samples contained D,L-2,3-butanediol in concentrations above 5.0 nmol/ml (range less than 5.0 nmol/ml-211 nmol/ml) and 7 (25%) of 29 samples contained meso-2,3-butanediol in concentrations above 5.0 nmol/ml (range less than 5.0-63.4 nmol/ml). In alcohol abstinent patients with alcoholic cirrhosis elevated serum 2,3-butanediol is due neither to ingestion of diols from undistilled alcoholic beverages nor is it likely to arise directly from the metabolic products of ethanol. In future studies on the origin of 2,3-butanediol in alcoholic patients biopsy evidence of the degree of associated liver damage should be obtained.
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Creatine, anhydrous
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
2,3-Butanediol, 98%