MilliporeSigma
  • Arginine infusion in patients with septic shock increases nitric oxide production without haemodynamic instability.
Arginine infusion in patients with septic shock increases nitric oxide production without haemodynamic instability.
Clinical science (London, England : 1979) (2014-07-19)
Yvette C Luiking, Martijn Poeze, Nicolaas E Deutz
ABSTRACT
Arginine deficiency in sepsis may impair nitric oxide (NO) production for local perfusion and add to the catabolic state. In contrast, excessive NO production has been related to global haemodynamic instability. Therefore, the aim of the present study was to investigate the dose-response effect of intravenous arginine supplementation in post-absorptive patients with septic shock on arginine-NO and protein metabolism and on global and regional haemodynamics. Eight critically ill patients with a diagnosis of septic shock participated in this short-term (8 h) dose-response study. L-Arginine-HCl was continuously infused [intravenously (IV)] in three stepwise-increasing doses (33, 66 and 99 μmol·kg-1·h-1). Whole-body arginine-NO and protein metabolism were measured using stable isotope techniques, and baseline values were compared with healthy controls. Global and regional haemodynamic parameters were continuously recorded during the study. Upon infusion, plasma arginine increased from 48±7 to 189±23 μmol·l-1 (means±S.D.; P<0.0001). This coincided with increased de novo arginine (P<0.0001) and increased NO production (P<0.05). Sepsis patients demonstrated elevated protein breakdown at baseline (P<0.001 compared with healthy controls), whereas protein breakdown and synthesis both decreased during arginine infusion (P<0.0001). Mean arterial and pulmonary pressure and gastric mucosal-arterial partial pressure of carbon dioxide difference (Pr-aCO2) gap did not alter during arginine infusion (P>0.05), whereas stroke volume (SV) increased (P<0.05) and arterial lactate decreased (P<0.05). In conclusion, a 4-fold increase in plasma arginine with intravenous arginine infusion in sepsis stimulates de novo arginine and NO production and reduces whole-body protein breakdown. These potential beneficial metabolic effects occurred without negative alterations in haemodynamic parameters, although improvement in regional perfusion could not be demonstrated in the eight patients with septic shock who were studied.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Millipore
Urea solution, suitable for microbiology, 40% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, suitable for electrophoresis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Citrulline, ≥98% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Arginine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
SKU
Pack Size
Availability
Price
Quantity
Arginine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Urea, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
USP
Urea, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Urea, European Pharmacopoeia (EP) Reference Standard