MilliporeSigma
Quantification of the effect of Lipo-PGE1 on angiogenesis.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS (2014-12-03)
SuRak Eo, Chan Kwon, HyunJic Lee, SangHun Cho, JinYoung Kim, GooHyun Baek, JeongSeok Yeo, ChiYeon Lim
ABSTRACT
Fifteen rabbits were used to assess the effect of Lipo-PGE1 on neovascularization. Merocel(®) and Alloderm(®) of the same size were implanted separately under the back skin to act as matrices for vessel growth. Lipo-PGE1 was injected intravenously for 2 weeks in an experimental group of eight rabbits, and they were compared with a control group of seven untreated animals. Blood flow was measured using the (99m)TcO4(-) clearance technique. The mean blood clearance halftime (T1/2) and washout radioactivity were measured. Newly formed vessels were counted by CD31. The mean clearance halftime was 4005 ± 2161.3 and 13840 ± 4644.6 s in the experimental and control group, respectively, in the 1 × 2 × 1.5-cm-sized implants (p = 0.0125), and 1560 ± 1174.7 and 3405 ± 807.03 s, respectively, in the 2 × 2 × 1.5-cm-sized implants (p = 0.0413). Histological examinations revealed that the mean numbers of newly formed vessels in the experimental and control groups were 11 ± 1.58 and 7.8 ± 1.71, respectively, in the 1 × 2 × 1.5-cm-sized implants (p = 0.0501), and 20.19 ± 12.47 and 12.33 ± 3.25, respectively, in the 2 × 2 × 1.5-cm-sized implants (p = 0.02679). Lipo-PGE1 was found to be effective in promoting angiogenesis in a rabbit matrix model.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Alprostadil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Poly(vinyl alcohol), SAJ first grade, degree of polymerization 500
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed