MilliporeSigma
  • Monoclonal antibodies against all known variants of EspA: development of a simple diagnostic test for enteropathogenic Escherichia coli based on a key virulence factor.
Monoclonal antibodies against all known variants of EspA: development of a simple diagnostic test for enteropathogenic Escherichia coli based on a key virulence factor.
Journal of medical microbiology (2014-09-19)
Uta Praekelt, Rolf Reissbrodt, Andreas Kresse, Asalapuram Pavankumar, Krishnan Sankaran, Roger James, Mary Jesudason, Shalini Anandan, Agila Prakasam, Veeraraghavan Balaji, Shanta Dutta, Sanjucta Dutta, Thandavarayan Ramamurthy, Renate Fischer, Peter Sander, Reiner Schaumann, Armando Navarro, Peter Williams
ABSTRACT
Enteropathogenic Escherichia coli (EPEC) are a major cause of infant diarrhoea in developing countries and a significant public health issue in industrialized countries. Currently there are no simple tests available for the diagnosis of EPEC. Serology of O-antigens is widely used routinely in many laboratories throughout the world, even though it has been known for many years to be an unreliable indicator of EPEC virulence. We have developed a simple, low-cost immunodiagnostic test based on the EspA filament, an essential virulence factor of EPEC and the related enterohaemorrhagic E. coli (EHEC). Using recombinant proteins of the five major variants of EspA as immunogens, we raised a panel of three monoclonal antibodies in mice that detects all variants of the native target in bacterial cultures. The antibodies proved suitable for application in sandwich-type assays, including ELISA and lateral flow immunoassays (LFI). Prototypes for both assays were specific for EPEC and EHEC strains when tested against a panel of control micro-organisms. We have also developed a simple, affordable culture medium, A/E medium, which optimizes expression of EspA allowing improved sensitivity of detection compared with standard Dulbecco's modified Eagle's medium. Together these reagents form the basis of robust, informative tests for EPEC for use especially in developing countries but also for routine screening in any clinical laboratory.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.