MilliporeSigma
  • Comparison of antibody molecules produced from two cell lines with contrasting productivities and aggregate contents.
Comparison of antibody molecules produced from two cell lines with contrasting productivities and aggregate contents.
Biological & pharmaceutical bulletin (2014-12-17)
Yoichi Ishii, Yasufumi Imamoto, Rie Yamamoto, Masayoshi Tsukahara, Kaori Wakamatsu
ABSTRACT
Cell culture processes that produce therapeutic antibodies with high productivity (titer) and low aggregate content reduce the risk of adverse effects and expense to patients. To elucidate the mechanism of aggregate formation, we compared trastuzumab samples produced from two contrasting cell lines: cell line A, which exhibits high titer and low aggregate content, and cell line B, which exhibits low titer and high aggregate content. Cell line B produced significantly fewer (approximately 1/3) antibodies compared with cell line A and contained higher (approximately 3-fold) percentages of aggregates. The aggregates of antibodies found in the protein A-purified samples of cell line B were associated mostly with noncovalent interactions. Cell line B exhibited a low content of monomers/dimers of light chains in the medium and within cells. Because light chains are essential for the correct folding of heavy chains and secretion of mature antibodies, the characteristics of cell line B may be attributed to low levels of light chain production. In addition, protein A-purified antibodies from cell line B (but not those from cell line A) contained fragments that are expected to expose the hydrophobic CH3 domain, which may serve as nuclei for aggregation.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Anti-Human IgG (γ-chain specific) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
D-Sorbitol, FCC, FG
SAFC
BIS-TRIS
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)