MilliporeSigma
  • Effect of muscle weakness and joint inflammation on the onset and progression of osteoarthritis in the rabbit knee.
Effect of muscle weakness and joint inflammation on the onset and progression of osteoarthritis in the rabbit knee.
Osteoarthritis and cartilage (2014-08-12)
C Egloff, A Sawatsky, T Leonard, D A Hart, V Valderrabano, W Herzog
ABSTRACT
Interactions between mechanical and non-mechanical independent risk factors for the onset and progression of Osteoarthritis (OA) are poorly understood. Therefore, the goal of the present study was to investigate the in vivo effects of muscle weakness, joint inflammation and the combination on the onset and progression of OA in a rabbit knee joint model. Thirty 1-year-old female New Zealand White rabbits (average 5.7 kg, range 4.8-6.6 kg) were divided into four groups with one limb randomly assigned to be the experimental side: (1) surgical denervation of the vastus lateralis (VL) muscle; (2) muscle weakness induced by intramuscular injection of Botulinum toxin A (BTX-A); (3) intraarticular injection with Carrageenan to induce a transient inflammatory reaction; (4) combination of Carrageenan and BTX-A injection. After 90 days, cartilage histology of the articular surfaces were microscopically analyzed using the Osteoarthritis Research Society International (OARSI) histology scoring system. VL denervation resulted in significantly higher OARSI scores in the patellofemoral joint (group 1). BTX-A administration resulted in significant cartilage damage in all four compartments of the knee (group 2). Carrageenan did not cause significant cartilage damage. BTX-A combined with Carrageenan lead to severe cartilage damage in all four compartments. Muscle weakness lead to significant OA in the rabbit knee. A transient local inflammatory stimulus did not promote cartilage degradation nor did it enhance OA progression when combined with muscle weakness. These results are surprising and add to the literature the conclusion that acute inflammation is probably not an independent risk factor for OA in this rabbit model.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%