MilliporeSigma
  • Effects of ceftriaxone on chronic ethanol consumption: a potential role for xCT and GLT1 modulation of glutamate levels in male P rats.
Effects of ceftriaxone on chronic ethanol consumption: a potential role for xCT and GLT1 modulation of glutamate levels in male P rats.
Journal of molecular neuroscience : MN (2014-02-19)
P S S Rao, Youssef Sari
ABSTRACT
Alterations in glutamatergic neurotransmission have been suggested to affect many aspects of neuroplasticity associated with alcohol/drug addiction. We have previously shown that ceftriaxone, a β-lactam antibiotic known to upregulate glutamate transporter 1 (GLT1), reduced ethanol intake after 5 weeks of free choice ethanol drinking paradigm in male alcohol-preferring (P) rats. Evidence suggests that differential effects involving alterations of glutamatergic neurotransmission occur after long-term ethanol consumption. In this study, we tested whether the efficacy of administration of ceftriaxone persists after 14 weeks of free access to 15 and 30 % ethanol in male P rats. After 14 weeks of ethanol consumption, male P rats were administered ceftriaxone (100 mg/kg, intraperitoneal (i.p.)) or saline vehicle for 5 days. We found that ceftriaxone treatment resulted in a significant reduction in ethanol intake starting from day 2 (48 h after the first i.p. injections of ceftriaxone) through day 14, 10 days after final injection. Western blot analysis of brain samples from animals euthanized 24 h after treatment with the last dose of ceftriaxone revealed a significant upregulation of cystine/glutamate exchanger (xCT) and GLT1 levels in prefrontal cortex, nucleus accumbens, and amygdala as compared to saline vehicle-treated group. These findings demonstrated the effectiveness of ceftriaxone in attenuating ethanol intake in a chronic consumption paradigm. These might be due in part through the upregulation of both xCT and GLT1 levels in brain reward regions. Thus, the drug has a potential therapeutic action for the treatment of alcohol dependence.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
Glycine
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard