Merck
  • Solid-state stability and compatibility studies of clavulanate potassium.
Solid-state stability and compatibility studies of clavulanate potassium.
Pharmaceutical development and technology (2013-11-14)
Judyta Cielecka-Piontek, Magdalena Paczkowska, Przemysław Zalewski, Kornelia Lewandowska, Bolesław Barszcz
ABSTRACT
The kinetic and thermodynamic parameters of degradation of clavulanate potassium in the solid state were studied by using a reversed phase high performance liquid chromatography (RP-HPLC) method. The degradation of clavulanate potassium was a first-order reaction depending on the substrate concentration at an increased relative air humidity (RH) and in dry air. The dependence ln k = f(1/T) became the ln k = (0.026 ± 166.35)-(2702.82 ± 1779.43)(1/T) in dry air and ln k = (1.65 ± 100.40) × 10(3)-(5748.81 ± 3659.67)(1/T) at 76.4% RH. The thermodynamic parameters Ea, ΔH(≠a), ΔS(≠a) of the degradation of clavulanate potassium in the solid state were calculated. The dependence ln k = f (RH%) assumed the form ln k = (8.78 ± 5.75) 10 (-2) (RH%) + (2.64 × 10(-8 )± 40.41). The compatibility of clavulanate potassium with commonly used excipients was studied at an increased temperature and in dry air. The geometric structure of molecule, highest occupied molecular orbital (HOMO) and lowest unoccupied molecular orbital (LUMO) orbitals were also determined in order to predict the structural changes and reactive sites in clavulanate potassium during degradation and compatibility studies in the solid state. The ultraviolet (UV), Fourier transform infrared spectroscopy (FT-IR) and Raman spectra of degraded samples of the compound were analyzed.
Product Description
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sigma-Aldrich
D-Lactose monohydrate, SAJ special grade
Supelco
Potassium clavulanate, VETRANAL®, analytical standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC)