Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.85%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, tablet
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sign Into View Organizational & Contract Pricing Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing SAFC
Sodium chloride solution, 5 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sign Into View Organizational & Contract Pricing Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in ethanol
Sign Into View Organizational & Contract Pricing