Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Supelco
2,2,2-Trifluoroethanol, analytical standard, for NMR spectroscopy
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, 99%
Sign Into View Organizational & Contract Pricing Supelco
L-Ascorbic acid, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sign Into View Organizational & Contract Pricing Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
E-Toxate™ Water, endotoxin, free
Sign Into View Organizational & Contract Pricing