Merck
  • Transplanted skin-derived precursor stem cells generate enteric ganglion-like structures in vivo.
Transplanted skin-derived precursor stem cells generate enteric ganglion-like structures in vivo.
Journal of pediatric surgery (2014-08-06)
Justin P Wagner, Veronica F Sullins, James C Y Dunn
ABSTRACT
Hirschsprung's disease is characterized by a developmental arrest of neural crest cell migration, causing distal aganglionosis. Transplanted cells derived from the neural crest may regenerate enteric ganglia in this condition. We investigated the potential of skin-derived precursor cells (SKPs) to engraft and to differentiate into enteric ganglia in aganglionic rat intestine in vivo. Adult Lewis rat jejunal segments were separated from intestinal continuity and treated with benzalkonium chloride to induce aganglionosis. Ganglia were identified via immunohistochemical stains for S100 and β-III tubulin (TUJ1). SKPs were procured from neonatal Lewis rats expressing enhanced green fluorescent protein (GFP) and cultured in neuroglial-selective media. SKP cell line expansion was quantified, and immunophenotypes were assessed by immunocytochemistry. Aganglionic segments underwent SKP transplantation 21-79days after benzalkonium chloride treatment. The presence of GFP+cells, mature neurons, and mature glia was evaluated at posttransplant days 1, 6, and 9. Benzalkonium chloride-induced aganglionosis persisted for at least 85days. Prior to differentiation, SKPs expressed S100, denoting neural crest lineage, and nestin, a marker of neuronal precursors. Differentiated SKPs in vitro expressed GFAP, a marker of glial differentiation, as well as TUJ1 and several enteric neurotransmitters. After transplantation, GFP+structures resembling ganglia were identified between longitudinal and circular smooth muscle layers. SKPs are capable of engraftment, migration, and differentiation within aganglionic rodent intestine in vivo. Differentiated SKPs generate structures that resemble enteric ganglia. Our observations suggest that SKPs represent a potential gangliogenic therapeutic agent for Hirschsprung's disease.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SKU
Pack Size
Availability
Price
Quantity
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, 10%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DAPI, for nucleic acid staining
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)