MilliporeSigma
  • Abundance of DLK1, differential expression of CYP11B1, CYP21A2 and MC2R, and lack of INSL3 distinguish testicular adrenal rest tumours from Leydig cell tumours.
Abundance of DLK1, differential expression of CYP11B1, CYP21A2 and MC2R, and lack of INSL3 distinguish testicular adrenal rest tumours from Leydig cell tumours.
European journal of endocrinology (2015-01-23)
Grete Lottrup, John E Nielsen, Niels E Skakkebæk, Anders Juul, Ewa Rajpert-De Meyts
ABSTRACT
Testicular adrenal rest tumours (TARTs) are a common finding in patients with congenital adrenal hyperplasia (CAH). These tumours constitute a diagnostic and management conundrum and may lead to infertility. TART cells share many functional and morphological similarities with Leydig cells (LCs), and masses consisting of such cells are occasionally misclassified as malignant testicular tumours, which may lead to erroneous orchiectomy in these patients. In this study, we aimed to investigate the potential of LC developmental markers and adrenal steroidogenic markers in the differential diagnosis of TARTs and malignant LC tumours (LCTs). We investigated mRNA and protein expression of testicular steroidogenic enzymes; CYP11A1 and HSD3B1/2, markers of adrenal steroidogenesis; CYP11B1, CYP21A2 and ACTH receptor/melanocortin 2 receptor (MC2R), and markers of LC maturation; and delta-like 1 homolog (DLK1) and insulin-like 3 (INSL3) in testicular biopsies with TART, orchiectomy specimens with LCTs and samples from human fetal adrenals. Expression of testicular steroidogenic enzymes was observed in all specimens. All investigated adrenal steroidogenic markers were expressed in TART, and weak reactions for CYP11B1 and MC2R were observed at the protein level in LTCs. TART and fetal adrenals had identical expression profiles. DLK1 was highly expressed and INSL3 not detectable in TART, whereas INSL3 was highly expressed in LCTs. The similar expression profiles in TART and fetal adrenals as well as the presence of classical markers of adrenal steroidogenesis lend support to the hypothesis that TART develops from a displaced adrenal cell type. Malignant LCTs seem to have lost DLK1 expression and do not resemble immature LCs. The different expression pattern of DLK1, INSL3 and most adrenocortical markers adds to the elucidation of the histogenesis of testicular interstitial tumours and may facilitate histopathological diagnosis.
Sigma-Aldrich
Anti-CYP11A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Anti-INSL3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)