MilliporeSigma
  • Sequential extraction procedure for the determination of some trace elements in fertilizer samples.
Sequential extraction procedure for the determination of some trace elements in fertilizer samples.
Journal of AOAC International (2014-08-26)
Yunus Emre Unsal, Mustafa Tuzen, Mustafa Soylak
ABSTRACT
Total concentration of metal ions at trace levels does not give sufficient information about toxicity and biological availability of these elements in fertilizer samples. In the presented work, a sequential extraction procedure modified by the European Community Bureau of Reference (BCR) was applied to fractionate Cd, Cr, Co, Cu, Fe, Pb, Mn, Ni, and Zn levels in two fertilizer samples collected from cooperative agricultural retailers. The fractions extracted were exchangeable/dilute acid soluble, reducible bound to Fe/Mn oxides, oxidizable bound to organic matter and sulfides, and residual. The determination of analyte elements was done by flame atomic absorption spectrometry. The accuracy of the procedure was validated with BCR-701 sediment certified reference material. The RSD of the procedure was less than 10%.
Sigma-Aldrich
Hydroxylamine hydrochloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer