MilliporeSigma
  • The biosurfactant viscosin transiently stimulates n-hexadecane mineralization by a bacterial consortium.
The biosurfactant viscosin transiently stimulates n-hexadecane mineralization by a bacterial consortium.
Applied microbiology and biotechnology (2014-09-14)
Frederik Bak, Lise Bonnichsen, Niels O G Jørgensen, Mette H Nicolaisen, Ole Nybroe
ABSTRACT
Pseudomonas produces powerful lipopeptide biosurfactants including viscosin, massetolide A, putisolvin, and amphisin, but their ability to stimulate alkane mineralization and their utility for bioremediation have received limited attention. The four Pseudomonas lipopeptides yielded emulsification indices on hexadecane of 20-31% at 90 mg/l, which is comparable to values for the synthetic surfactant Tween 80. Viscosin was the optimal emulsifier and significantly stimulated n-hexadecane mineralization by diesel-degrading bacterial consortia but exclusively during the first 2 days of batch culture experiments. Growth of the consortia, as determined by OD₆₀₀ measurements and quantification of the alkB marker gene for alkane degradation, was arrested after the first day of the experiment. In contrast, the control consortia continued to grow and reached higher OD₆₀₀ values and higher alkB copy numbers during the next days. Due to the short-lived stimulation of n-hexadecane mineralization, the stability of viscosin was analyzed, and it was observed that added viscosin was degraded by the bacterial consortium during the first 2 days. Hence, viscosin has a potential as stimulator of alkane degradation, but its utility in bioremediation may be limited by its rapid degradation and growth-inhibiting properties.
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Supelco
Hexadecane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Pricing and availability is not currently available.
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Pricing and availability is not currently available.
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Pricing and availability is not currently available.