MilliporeSigma
  • Time course study of the antigen-specific immune response to a PLGA microparticle vaccine formulation.
Time course study of the antigen-specific immune response to a PLGA microparticle vaccine formulation.
Biomaterials (2014-07-06)
Qian Wang, Melody T Tan, Brian P Keegan, Meagan A Barry, Michael J Heffernan
ABSTRACT
Microparticle-based vaccine delivery systems are known to promote enhanced immune responses to protein antigens and can elicit TH1-biased responses when used in combination with Toll-like receptor (TLR) agonists. It is important to understand the kinetics of the immune responses to microparticle-based protein vaccines in order to predict the duration of protective immunity and to optimize prime-boost vaccination regimens. We carried out a 10-week time course study to investigate the magnitude and kinetics of the antibody and cellular immune responses to poly(lactic-co-glycolic acid) (PLGA) microparticles containing 40 μg ovalbumin (OVA) protein and 16 μg CpG-ODN adjuvant (MP/OVA/CpG) in comparison to OVA-containing microparticles, soluble OVA plus CpG, or OVA formulated with Alhydrogel(®) aluminum adjuvant. Mice vaccinated with MP/OVA/CpG developed the highest TH1-associated IgG2b and IgG2c antibody titers, while also eliciting TH2-associated IgG1 antibody titers on par with Alhydrogel(®)-formulated OVA, with all IgG subtype titers peaking at day 56. The MP/OVA/CpG vaccine also induced the highest antigen-specific splenocyte IFN-γ responses, with high levels of IFN-γ responses persisting until day 42. Thus the MP/OVA/CpG formulation produced a sustained and heightened humoral and cellular immune response, with an overall TH1 bias, while maintaining high levels of IgG1 antibody equivalent to that seen with Alhydrogel(®) adjuvant. The time course kinetics study provides a useful baseline for designing vaccination regimens for microparticle-based protein vaccines.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
Dichloromethane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Supelco
Dichloromethane, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μL
Estimated to ship on March 21, 2025
$411.00