MilliporeSigma
  • Enhanced delivery of artemisinin and its analogues to cancer cells by their adducts with human serum transferrin.
Enhanced delivery of artemisinin and its analogues to cancer cells by their adducts with human serum transferrin.
International journal of pharmaceutics (2014-03-26)
Yan Yang, Xiaomin Zhang, Xiaofen Wang, Xiaomei Zhao, Tianrui Ren, Feng Wang, Bo Yu
ABSTRACT
Artemisinin (ART) and its analogues, such as dihydroartesunate (DHA) and artesunate (ATS), are sesquiterpene lactones with anticancer activities. Transferrin (Tf) receptor is frequently overexpressed in cancer cells. In order to improve the delivery and the anticancer activity of ART and its analogues, adducts of Tf with ART, DHA or ATS were fabricated by simply combining ART, DHA or ATS with Tf. Increased antitumor effects of these adducts were observed on human liver hepatocellular carcinoma (HepG2) and lung adenocarcinoma (A549) cells. Meanwhile, only a low level of toxic effect was observed on normal human liver cells (HL-7702). Improved cellular uptake of ATS-Tf adduct compared to ATS alone was confirmed by HPLC analysis. UV-vis, fluorescence spectroscopy and docking study further confirmed the formation of the adducts with relatively high binding constants at neutral pH (7.4×10(4), 4.2×10(5) and 3.4×10(5)M(-1), for ART-Tf, DHA-Tf and ATS-Tf, respectively, at pH 7.4). However, the adducts became less stable with reduced binding constants under an acidic condition (2.6×10(4), 1.9×10(4) and 1.7×10(4)M(-1), for ART-Tf, DHA-Tf and ATS-Tf, respectively, at pH 5.5). A possible mechanism of the anticancer effect by these adducts was proposed. The short term and long term stability of ART-Tf in the presence of human serum albumin (HSA) was also studied. Our results showed that adducts of ART and its analogues with Tf, especially ATS-Tf and DHA-Tf, have significant anticancer effects to cancer cells, with minimal side effects on normal cells, therefore, are promising as potential novel anticancer agents.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid solution, suitable for HPLC