MilliporeSigma
  • Trefoil factor 3 (TFF3) enhances the oncogenic characteristics of prostate carcinoma cells and reduces sensitivity to ionising radiation.
Trefoil factor 3 (TFF3) enhances the oncogenic characteristics of prostate carcinoma cells and reduces sensitivity to ionising radiation.
Cancer letters (2015-03-10)
Omesha Perera, Angharad Evans, Mikhail Pertziger, Christa MacDonald, Helen Chen, Dong-Xu Liu, Peter E Lobie, Jo K Perry
ABSTRACT
Trefoil factor 3 (TFF3) is a secreted protein which functions in mucosal repair of the gastrointestinal tract. This is achieved through the combined stimulation of cell migration and prevention of apoptosis and anoikis, thus facilitating repair. Deregulated TFF3 expression at the gene and protein level is implicated in numerous cancers. In prostate cancer TFF3 has previously been reported as a potential biomarker, overexpressed in a subset of primary and metastatic cases. Here we investigated the effect of increased TFF3 expression on prostate cancer cell behaviour. Oncomine analysis demonstrated that TFF3 mRNA expression was upregulated in prostate cancer compared to normal tissue. Forced-expression models were established in the prostate cancer cell lines, DU145 and PC3, by stable transfection of an expression vector containing the TFF3 cDNA. Forced expression of TFF3 significantly increased total cell number and cell viability, cell proliferation and cell survival. In addition, TFF3 enhanced anchorage independent growth, 3-dimensional colony formation, wound healing and cell migration compared to control transfected cell lines. We also observed reduced sensitivity to ionising radiation in stably transfected cell lines. In dose response experiments, forced expression of TFF3 significantly enhanced the regrowth of PC3 cells following ionising radiation compared with control transfected cells. In addition, TFF3 enhanced clonogenic survival of DU145 and PC3 cells. These studies indicate that targeting TFF3 for the treatment of prostate cancer warrants further investigation.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%