MilliporeSigma
  • Ferrocenyl-L-amino acid copper(II) complexes showing remarkable photo-induced anticancer activity in visible light.
Ferrocenyl-L-amino acid copper(II) complexes showing remarkable photo-induced anticancer activity in visible light.
Dalton transactions (Cambridge, England : 2003) (2014-06-28)
Tridib K Goswami, Sudarshan Gadadhar, Babu Balaji, Bappaditya Gole, Anjali A Karande, Akhil R Chakravarty
ABSTRACT
Ferrocene-conjugated copper(ii) complexes [Cu(Fc-aa)(aip)](ClO4) () and [Cu(Fc-aa)(pyip)](ClO4) () of l-amino acid reduced Schiff bases (Fc-aa), 2-(9-anthryl)-1H-imidazo[4,5-f][1,10]phenanthroline (aip) and 2-(1-pyrenyl)-1H-imidazo[4,5-f][1,10]phenanthroline (pyip), where Fc-aa is ferrocenylmethyl-l-tyrosine (Fc-Tyr in , ), ferrocenylmethyl-l-tryptophan (Fc-Trp in , ) and ferrocenylmethyl-l-methionine (Fc-Met in , ), were prepared and characterized, and their photocytotoxicity was studied (Fc = ferrocenyl moiety). Phenyl analogues, viz. [Cu(Ph-Met)(aip)](ClO4) () and [Cu(Ph-Met)(pyip)](ClO4) (), were prepared and used as control compounds. The bis-imidazophenanthroline copper(ii) complexes, viz. [Cu(aip)2(NO3)](NO3) () and [Cu(pyip)2(NO3)](NO3) (), were also prepared and used as controls. Complexes having a redox inactive cooper(ii) center showed the Fc(+)-Fc redox couple at ∼0.5 V vs. SCE in DMF-0.1 mol [Bu(n)4N](ClO4). The copper(ii)-based d-d band was observed near 600 nm in DMF-Tris-HCl buffer (1 : 1 v/v). The ferrocenyl complexes showed low dark toxicity, but remarkably high photocytotoxicity in human cervical HeLa and human breast adenocarcinoma MCF-7 cancer cells giving an excellent photo-dynamic effect while their phenyl analogues were inactive. The photo-exposure caused significant morphological changes in the cancer cells when compared to the non-irradiated ones. The photophysical processes were rationalized from the theoretical studies. Fluorescence microscopic images showed and localizing predominantly in the endoplasmic reticulum (ER) of the cancer cells, thus minimizing any undesirable effects involving nuclear DNA.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
9-Anthracenecarboxaldehyde, 97%
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%