MilliporeSigma
  • Inhibition of STAT5a by Naa10p contributes to decreased breast cancer metastasis.
Inhibition of STAT5a by Naa10p contributes to decreased breast cancer metastasis.
Carcinogenesis (2014-06-14)
Yan Zeng, Li Min, Yong Han, Lin Meng, Caiyun Liu, Yuntao Xie, Bin Dong, Lixin Wang, Beihai Jiang, Huiyu Xu, Qing Zhuang, Chuanke Zhao, Like Qu, Chengchao Shou
ABSTRACT
N-α-Acetyltransferase 10 protein (Naa10p, also called arrest-defective 1), the catalytic subunit of N-acetyltransferase A, is a critical regulator of cell death and proliferation. Naa10p is also shown to regulate cancer metastasis by inhibiting cell motility; however, its role in cancer metastasis is not fully understood. In this study, we found that high expression of Naa10p is positively correlated with the survival of patients with breast cancer, whereas negatively correlated with lymph node metastasis. Naa10p inhibits breast cancer cell migration and invasion in vitro and decreases the xenograft growth and metastasis in nude mice. Microarray screening revealed that Naa10p downregulates inhibitors of differentiation 1 (ID1) expression. Naa10p binds to signal transducer and activator of transcription 5a (STAT5a) and decreases STAT5a-stimulated ID1 expression in an acetyltransferase-independent manner. Moreover, Naa10p antagonizes Janus kinase 2-STAT5a signaling by lowering p65-activated interleukin-1β expression. Our results demonstrate a novel mechanism through which Naa10p inhibits the metastasis of breast cancer cells by targeting STAT5a.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)