MilliporeSigma
  • Interleukin (IL)-22 receptor 1 is over-expressed in primary Sjogren's syndrome and Sjögren-associated non-Hodgkin lymphomas and is regulated by IL-18.
Interleukin (IL)-22 receptor 1 is over-expressed in primary Sjogren's syndrome and Sjögren-associated non-Hodgkin lymphomas and is regulated by IL-18.
Clinical and experimental immunology (2015-04-17)
F Ciccia, G Guggino, A Rizzo, M Bombardieri, S Raimondo, F Carubbi, A Cannizzaro, G Sireci, F Dieli, G Campisi, R Giacomelli, Paola Cipriani, G De Leo, R Alessandro, G Triolo
ABSTRACT
The aim of this study was to elucidate more clearly the role of interleukin (IL)-18 in modulating the IL-22 pathway in primary Sjögren's syndrome (pSS) patients and in pSS-associated lymphomas. Minor salivary glands (MSGs) from patients with pSS and non-specific chronic sialoadenitis (nSCS), parotid glands biopsies from non-Hodgkin lymphomas (NHL) developed in pSS patients, were evaluated for IL-18, IL-22, IL-22 receptor 1 (IL-22R1), IL-22 binding protein (IL-22BP) and signal transducer and activator of transcription-3 (STAT-3) expression. MSGs IL-22R1-expressing cells were characterized by confocal microscopy and flow cytometry in pSS, nSCS and healthy controls . The effect of recombinant IL-18 and IL-22 on peripheral blood mononuclear cells (PBMCs) from pSS and nSCS was studied by flow cytometry and reverse transcription-polymerase chain reaction (RT-PCR). MSGs of pSS and NHL were characterized by an imbalance between IL-22 and IL-22BP protein expression, with IL-18 and IL-22BP being expressed in a mutually exclusive manner and IL-18 and IL-22R1 being correlated directly. Aberrant expression of IL-22R1, induced by IL-18, was observed only among tissue and circulating myeloid cells of pSS patients and macrophages of NHL tissues of pSS patients, but not nSCS. IL-22R1 expression on PBMC of pSS was functional, as its stimulation with recombinant IL-22 significantly up-regulated the expression of STAT-3, IL-17 and IL-22. An IL-18-dependent aberrant expression of IL-22R1 on cells of haematopoietic origin seems to be a specific immunological signature of patients with pSS and pSS-associated lymphomas.
Sigma-Aldrich
IL-22 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
SAFC
HEPES
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
IL-22 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
IL-22 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
SAFC
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 24, 2025
$53.20
5 g
Estimated to ship on March 24, 2025
$208.00
10 g
Estimated to ship on March 24, 2025
$431.00